Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry 2-Methyl-1-tetralone | 2G | 1590-08-5 | MFCD00001690

Catalog No. 16322825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

CAS #: 1590-08-5
MDL #: MFCD00001690
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 160.21

Catalog No. 16-322-825G Supplier Sigma Organic Chemistry Supplier No. 1632282.5G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #: 1590-08-5
MDL #: MFCD00001690
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 160.21

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.