Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Catalog No. SB01361EA
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SB01361EA 10 g Amber Glass Bottle
SB01361DA 1 g Amber Glass Bottle
SB01361EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SB01361EA Supplier Thermo Scientific Chemicals Supplier No. SB01361EA
Add to Cart

Chemical Identifiers

CAS 35034-22-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
InChI Key ZWULFIBGPXWGFG-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbaldehyde, 2-methyl-4-formylimidazole, 2-methylimidazole-4-carbaldehyde, 4-formyl-2-methyl-1h-imidazole, 2-methylimidazole-5-carboxaldehyde, 2-methyl-3h-imidazole-4-carbaldehyde, 2-methyl-1h-imidazole-4-carboxaldehyde, 4-formyl-2-methylimidazole, 2-methyl-4-imidazolecarboxaldehyde, 2-methyl-1h-imidazole-5-carboxaldehyde
PubChem CID 2795878
IUPAC Name 2-methyl-1H-imidazole-5-carbaldehyde
SMILES CC1=NC=C(N1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 110.12
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.