Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-adamantanol 99.0+%, TCI America™
SDP

Catalog No. M13345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13345G 5 g
M133425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13345G Supplier TCI America Supplier No. M13345G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 702-98-7
Molecular Formula C11H18O
Molecular Weight (g/mol) 166.26
MDL Number MFCD00074811
InChI Key JKOZWMQUOWYZAB-UHFFFAOYSA-N
Synonym 2-methyl-2-adamantanol, 2-adamantanol, 2-methyl, tricyclo 3.3.1.13,7 decan-2-ol, 2-methyl, 1r,3r,5r,7r-2-methyladamantan-2-ol, tricyclo 3.3.1.1 3,7 decan-2-ol, 2-methyl, pubchem8767, 2-methyl-2-adamantol, acmc-1bihj, maybridge1_004158, 2-hydroxy-2-methyladamantane
PubChem CID 136545
IUPAC Name 2-methyladamantan-2-ol
SMILES CC1(O)C2CC3CC(C2)CC1C3

Specifications

Melting Point 217°C
Color White
Quantity 5 g
Formula Weight 166.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2-adamantanol
EINECSNumber (4)-1929
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.