Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Methyl-2-Butanol

Catalog No. 2404865ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-048-65ML 5 mL
24-048-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 24-048-65ML Supplier Sigma Aldrich Supplier No. 2404865ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Methyl-2-Butanol

Specifications

CAS 75-85-4
Melting Point -12°C
Density 0.805 g/mL at 25°C
Boiling Point 102°C
Molecular Formula C5H12O
Refractive Index n20/D 1.405
Linear Formula CH3CH2C(CH3)2OH
MDL Number MFCD00004478
Quantity 5 mL
Synonym tert-Amyl alcohol; tert-Pentyl alcohol
Molecular Weight (g/mol) 88.15 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.