Learn More
Sigma Aldrich Fine Chemicals Biosciences 2-Methyl-2-butanol Reagent
Catalog No. 502899365
Encompass
2-Methyl-2-butanol is an isomeric form of pentanol. Various physical properties (static dielectric constant viscosity density ultrasonic and dielectric relaxation) of 2-methyl-2-butanol have been evaluated over a wide range of temperatures. Its ability as solvent in the extraction of furfural from aqueous SOLUTIONons has been tested. Kinetic parameters of the three-phase reaction of MBY with hydrogen catalyzed by a commercial palladium-based catalyst in the absence of solvent have been described.
Catalog No. 50-289-9365 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1524632.5L
2-Methyl-2-butanol is an isomeric form of pentanol. Various physical properties (static dielectric constant viscosity density ultrasonic and dielectric relaxation) of 2-methyl-2-butanol have been evaluated over a wide range of temperatures. Its ability as solvent in the extraction of furfural from aqueous SOLUTIONons has been tested. Kinetic parameters of the three-phase reaction of MBY with hydrogen catalyzed by a commercial palladium-based catalyst in the absence of solvent have been described.