Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-butene, ≥99% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019919 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9919 5 mL
11-101-9097 10 mL
11-101-8191 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-9919 Supplier MilliporeSigma Supelco Supplier No. 660505ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-2-butene is a trisubstituted olefin which is mostly used as a precursor for peroxyacetyl nitrate. It can be prepared by the acid-catalyzed dehydration of 3-methyl-2-butanol. It has 3 alkyl groups on the double bond.

Featured Industries: Environmental and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 513-35-9
Vapor Density 2.4 (vs. air)
Density 0.662 g/mL (at 25°C)
Flash Point -20°C
Physical Form Neat
Linear Formula CH3 CH=C(CH3)2
MDL Number MFCD00009276
Melting Point -134°C
Boiling Point 35°C to 38°C
Synonym beta-Isoamylene; Amylene
Type Analytical Standard
Quantity 5 mL
Formula Weight 70.13
Percent Purity ≥99% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.