Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-nitro-1,3-propanediol, 97%

Catalog No. AAB2519606
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2519606

Supplier: Thermo Scientific Chemicals B2519606

Only null left
Add to Cart

CAS: 77-49-6 | C4H9NO4 | 135.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-49-6
Molecular Formula C4H9NO4
Molecular Weight (g/mol) 135.12
MDL Number MFCD00024812
InChI Key LOTYADDQWWVBDJ-UHFFFAOYSA-N
Synonym 2-methyl-2-nitro-1,3-propanediol, nmpd, 2-nitro-2-methyl-1,3-propanediol, nitroisobutylglycol, 1,3-propanediol, 2-methyl-2-nitro, 2-methyl-2-nitropropanediol, 1,1-dimethylol-1-nitroethane, caswell no. 573n, epa pesticide chemical code 573900, acmc-1basy
PubChem CID 6480
IUPAC Name 2-methyl-2-nitropropane-1,3-diol
SMILES CC(CO)(CO)[N+]([O-])=O

Specifications

Melting Point 148°C to 151°C
Quantity 5 g
Formula Weight 135.12
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-nitro-1,3-propanediol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-031-4
RTECSNumber TY8925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.