Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-nitrosopropane dimer, 98%, Thermo Scientific™

Catalog No. AC177342500
Change view
Click to view available options
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC177342500 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC177342500 Supplier Thermo Scientific Chemicals Supplier No. 177342500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6841-96-9
CAS Min % 97.5
CAS Max % 100.0
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C8H18N2O2
Linear Formula {(CH3)3CNO}2
Quantity 250mg
MDL Number MFCD00002065
Beilstein 01, 129
Synonym dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide
InChI Key PKKJRWITDTTZCL-UHFFFAOYSA-N
SMILES CC(C)(C)N=O.CC(C)(C)N=O
IUPAC Name 2-methyl-2-nitrosopropane
Molecular Weight (g/mol) 174.244
PubChem CID 44134826
Formula Weight 174.24
Chemical Name or Material 2-Methyl-2-nitrosopropane dimer, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.