Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Methyl-2-oxazoline, 99%

Click to view available options
Quantity:
25g
100g
1120-64-5
C4H7NO
85.106
MFCD00005298
GUXJXWKCUUWCLX-UHFFFAOYSA-N
2-methyl-2-oxazoline,2-methyloxazoline,2-methyl-4,5-dihydrooxazole,oxazole, 4,5-dihydro-2-methyl,2-methyl-2-oxazolin,2-methyl-1,3-oxazoline,2-oxazoline, 2-methyl,4,5-dihydro-2-methyl-1,3-oxazole,poly 2-methyl-2-oxazoline,methyl oxazoline
70713
CHEBI:53614
2-methyl-4,5-dihydro-1,3-oxazole
CC1=NCCO1
1120-64-5
C4H7NO
85.106
MFCD00005298
GUXJXWKCUUWCLX-UHFFFAOYSA-N
2-methyl-2-oxazoline,2-methyloxazoline,2-methyl-4,5-dihydrooxazole,oxazole, 4,5-dihydro-2-methyl,2-methyl-2-oxazolin,2-methyl-1,3-oxazoline,2-oxazoline, 2-methyl,4,5-dihydro-2-methyl-1,3-oxazole,poly 2-methyl-2-oxazoline,methyl oxazoline
70713
CHEBI:53614
2-methyl-4,5-dihydro-1,3-oxazole
CC1=NCCO1
Applications
2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N′-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

Solubility
Soluble in water (7051 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Quantity 25g
Flash Point 20°C (68°F)
Boiling Point 109°C to 111°C
UN Number UN1993
Beilstein 104227
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-oxazoline

GHS H Statement
H225
Highly flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.