Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-pentenoic acid, 99%

Catalog No. AAA1658709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1658709 10 g
AAA1658718 50 g
AAA1658730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1658709 Supplier Thermo Scientific Chemicals Supplier No. A1658709
Add to Cart

CAS: 3142-72-1 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3142-72-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O

Specifications

Density 0.978
Boiling Point 120°C (20 mmHg)
Flash Point 107°C (225°F)
Odor Characteristic
Quantity 10 g
UN Number UN3265
Formula Weight 114.15
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentenoic acid
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 221-552-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.