Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2-Methyl-2-propanesulfinamide, 97%

Catalog No. AAB2538503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2538503 1 g
AAB2538506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2538503 Supplier Thermo Scientific Chemicals Supplier No. B2538503
Add to Cart

CAS: 146374-27-8 | C4H11NOS | 121.20 g/mol

(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146374-27-8
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.20
MDL Number MFCD01863616,MFCD05861480
InChI Key CESUXLKAADQNTB-UHFFFAOYNA-N
Synonym 2-methyl-2-propanesulfinamide, tert-butanesulfinamide, t-butylsulfinamide, tert-butylsulfinamide, 1,1-dimethylethylsulfinamide, 2-propanesulfinamide, 2-methyl, 2-methyl-propane-2-sulfinic acid amide, tert-butylsulphinamide, unii-i85yc2za8o, i85yc2za8o
PubChem CID 3382465
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(N)=O

Specifications

Melting Point 97°C to 101°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 121.2
Percent Purity 97%
Chemical Name or Material (±)-2-Methyl-2-propanesulfinamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.