Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methyl-2-thiazoline-4-carboxylic acid (sodium) | 15058-19-2 | MFCD03093520 | 98.1% | 167.16 | C5H6NNaO2S | 50 MG
SDP

Catalog No. 5000282769
Encompass_Preferred

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline carboxylic acid supplied as a solid research reagent. It is used as a heterocyclic building block and biochemical reagent in organic synthesis and laboratory studies that require a thiazoline-derived carboxylate.

  • Solid, light yellow to yellow appearance.
  • Purity 98.1%.
  • Molecular weight 167.16 g/mol; formula C5H6NNaO2S.
  • Identified by CAS number 15058-19-2.
  • Available in small-scale laboratory quantities (25 mg-10 g).
  • Suitable as a heterocyclic building block and biochemical reagent for organic synthesis and research.

Catalog No. 50-002-82769 Supplier Medchemexpress LLC Supplier No. HYW14035150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline carboxylic acid supplied as a solid research reagent. It is used as a heterocyclic building block and biochemical reagent in organic synthesis and laboratory studies that require a thiazoline-derived carboxylate.

  • Solid, light yellow to yellow appearance.
  • Purity 98.1%.
  • Molecular weight 167.16 g/mol; formula C5H6NNaO2S.
  • Identified by CAS number 15058-19-2.
  • Available in small-scale laboratory quantities (25 mg-10 g).
  • Suitable as a heterocyclic building block and biochemical reagent for organic synthesis and research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.