Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methyl-2-thiazoline-4-carboxylic acid (sodium) | 15058-19-2 | MFCD03093520 | 98.1% | 167.16 g/mol | C5H6NNaO2S | 1 G
SDP

Catalog No. 5000283600
Encompass_Preferred

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline-class heterocycle supplied as a solid research reagent. It is used as a synthetic building block and biochemical intermediate for laboratory research. Consult the SDS for handling, storage, and safety information.

  • High purity: 98.1% (typical).
  • Molecular weight: 167.16 g/mol.
  • Chemical formula: C5H6NNaO2S.
  • CAS number: 15058-19-2.
  • Physical form: sodium salt, solid.
  • Common applications: synthetic building block and biochemical research.
  • Documentation: SDS and datasheet available for handling and safety.

Catalog No. 50-002-83600 Supplier Medchemexpress LLC Supplier No. HYW1403511G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline-class heterocycle supplied as a solid research reagent. It is used as a synthetic building block and biochemical intermediate for laboratory research. Consult the SDS for handling, storage, and safety information.

  • High purity: 98.1% (typical).
  • Molecular weight: 167.16 g/mol.
  • Chemical formula: C5H6NNaO2S.
  • CAS number: 15058-19-2.
  • Physical form: sodium salt, solid.
  • Common applications: synthetic building block and biochemical research.
  • Documentation: SDS and datasheet available for handling and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.