Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methyl-2-thiazoline-4-carboxylic acid (sodium) | 15058-19-2 | MFCD03093520 | 98.1% | 167.16 g/mol | C5H6NNaO2S | 100 MG
SDP

Catalog No. 5000282712
Encompass_Preferred

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of 2-methyl-2-thiazoline-4-carboxylic acid supplied as a purified research reagent for laboratory use. It is suitable for biochemical and medicinal chemistry research involving thiazoline derivatives and is offered in small pack sizes for bench-top experiments.

  • Purity 98.1%.
  • Chemical formula C5H6NNaO2S.
  • Molecular weight 167.16 g/mol.
  • CAS number 15058-19-2.
  • Available in small pack sizes including 100 MG.

Catalog No. 50-002-82712 Supplier Medchemexpress LLC Supplier No. HYW140351100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of 2-methyl-2-thiazoline-4-carboxylic acid supplied as a purified research reagent for laboratory use. It is suitable for biochemical and medicinal chemistry research involving thiazoline derivatives and is offered in small pack sizes for bench-top experiments.

  • Purity 98.1%.
  • Chemical formula C5H6NNaO2S.
  • Molecular weight 167.16 g/mol.
  • CAS number 15058-19-2.
  • Available in small pack sizes including 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.