Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-methyl-2-thiazoline-4-carboxylic acid sodium | 15058-19-2 | MFCD03093520 | 98.1% | 167.16 g/mol | C5H6NNaO2S | 500 MG
SDP

Catalog No. 5000283566
Encompass_Preferred

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline-class organic acid supplied for research use. It is a light yellow to yellow solid with formula C5H6NNaO2S and molecular weight 167.16 g/mol. The material is provided with high purity and standard laboratory storage and solubility characteristics.

  • Purity 98.07%.
  • Molecular formula C5H6NNaO2S.
  • Molecular weight 167.16 g/mol.
  • Appearance light yellow to yellow solid.
  • Solubility in water 50 mg/mL (requires ultrasonic).
  • Storage -20°C, sealed, protect from moisture and light.
  • CAS number 15058-19-2.

Catalog No. 50-002-83566 Supplier Medchemexpress LLC Supplier No. HYW140351500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-2-thiazoline-4-carboxylic acid sodium is the sodium salt of a thiazoline-class organic acid supplied for research use. It is a light yellow to yellow solid with formula C5H6NNaO2S and molecular weight 167.16 g/mol. The material is provided with high purity and standard laboratory storage and solubility characteristics.

  • Purity 98.07%.
  • Molecular formula C5H6NNaO2S.
  • Molecular weight 167.16 g/mol.
  • Appearance light yellow to yellow solid.
  • Solubility in water 50 mg/mL (requires ultrasonic).
  • Storage -20°C, sealed, protect from moisture and light.
  • CAS number 15058-19-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.