Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-2-thiazoline, 97%

Catalog No. AAL0132922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0132922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0132922 Supplier Thermo Scientific Chemicals Supplier No. L0132922
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2346-00-1 | C4H7NS | 101.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2346-00-1
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.17
MDL Number MFCD00005314
InChI Key JUIQOABNSLTJSW-UHFFFAOYSA-N
Synonym 2-methyl-2-thiazoline, 2-methylthiazoline, thiazole, 4,5-dihydro-2-methyl, 2-methyl-4,5-dihydrothiazole, 4,5-dihydro-2-methylthiazole, methyl-2-thiazoline, 2-thiazoline, 2-methyl, unii-8z6ua8v7wn, methyl-2 delta-2 thiazoline, methyl-2 delta-2 thiazoline french
PubChem CID 16867
IUPAC Name 2-methyl-4,5-dihydro-1,3-thiazole
SMILES CC1=NCCS1

Specifications

Melting Point -101°C
Density 1.072
Boiling Point 144°C to 145°C
Flash Point 37°C (98°F)
Refractive Index 1.52
Quantity 100 g
UN Number UN1993
Beilstein 104274
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-thiazoline
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-071-6
RTECSNumber XJ4261470
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.