Learn More
Medchemexpress LLC 2-methyl-3-[(2e,7r,11r)-3,7,11,15-tetramethylhexadec-2-en-1-yl]naphthalene-1,4-dione | 84-80-0 | MFCD00075678 | 450.70 | 50 MG
SDP
Catalog No. 5000416674
Encompass_Preferred
Vitamin K1 (phylloquinone) is a naturally occurring fat-soluble vitamin crucial for blood coagulation, bone metabolism, and vascular health. It acts as a cofactor for gamma-glutamyl carboxylase, essential for modifying proteins involved in these processes.
  • Blood coagulation: essential for synthesizing coagulation factors in the liver.
  • Bone metabolism: involved in osteocalcin carboxylation for bone mineralization.
  • Vascular health: promotes matrix Gla protein (MGP) carboxylation, preventing arterial hardening.
  • Antioxidant properties: may protect cells from oxidative damage.
Catalog No. 50-004-16674 Supplier Medchemexpress LLC Supplier No. HYN068450G
Vitamin K1 (phylloquinone) is a naturally occurring fat-soluble vitamin crucial for blood coagulation, bone metabolism, and vascular health. It acts as a cofactor for gamma-glutamyl carboxylase, essential for modifying proteins involved in these processes.
  • Blood coagulation: essential for synthesizing coagulation factors in the liver.
  • Bone metabolism: involved in osteocalcin carboxylation for bone mineralization.
  • Vascular health: promotes matrix Gla protein (MGP) carboxylation, preventing arterial hardening.
  • Antioxidant properties: may protect cells from oxidative damage.