Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Tokyo Chemical Industry, 2-Methyl-3-butyn-2-ol, Reagent, 98.0%, 25 mL

Catalog No. 502781463
Encompass_Preferred

2-Methyl-3-butyn-2-ol is a chemical reagent typically used in laboratory research settings. This terminal alkyne acts as a versatile building block in chemical synthesis, particularly in ethynylation reactions and conjugation chemistry.

  • Purity: ≥98.0% (GC)
  • Molecular formula: C5H8O
  • Molecular weight: 84.12 g/mol
  • Physical state: Clear liquid
  • Boiling point: 103 °C
  • Flash point: 19 °C
  • Water solubility: Completely miscible
  • Storage temperature: Room temperature (recommended <15 °C)

Catalog No. 50-278-1463 Supplier Grainger Supplier No. 19KU52
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-3-butyn-2-ol is a chemical reagent typically used in laboratory research settings. This terminal alkyne acts as a versatile building block in chemical synthesis, particularly in ethynylation reactions and conjugation chemistry.

  • Purity: ≥98.0% (GC)
  • Molecular formula: C5H8O
  • Molecular weight: 84.12 g/mol
  • Physical state: Clear liquid
  • Boiling point: 103 °C
  • Flash point: 19 °C
  • Water solubility: Completely miscible
  • Storage temperature: Room temperature (recommended <15 °C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.