Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger TCI AMERICA 2-Methyl-3-butyn-2-ol, >98.0% Purity, Liquid

Catalog No. 502780943
Encompass_Preferred

2-Methyl-3-butyn-2-ol is a clear, colorless to light yellow liquid utilized as a laboratory reagent. It features an acetylenic functional group, making it suitable for applications in chemical synthesis, ethynylation, and conjugation chemistry.

  • Purity of 98.0% or greater by gas chromatography
  • Molecular formula of C5H8O and molecular weight of 84.12 g/mol
  • Physical state as a liquid at 20°C
  • Completely miscible in water
  • Stable under normal conditions
  • Recommended storage at room temperature in a cool, dark place (below 15°C)

Catalog No. 50-278-0943 Supplier Grainger Supplier No. 19KU53
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methyl-3-butyn-2-ol is a clear, colorless to light yellow liquid utilized as a laboratory reagent. It features an acetylenic functional group, making it suitable for applications in chemical synthesis, ethynylation, and conjugation chemistry.

  • Purity of 98.0% or greater by gas chromatography
  • Molecular formula of C5H8O and molecular weight of 84.12 g/mol
  • Physical state as a liquid at 20°C
  • Completely miscible in water
  • Stable under normal conditions
  • Recommended storage at room temperature in a cool, dark place (below 15°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.