Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-(methylthio)furan 98.0+%, TCI America™
SDP

Catalog No. M18385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18385G 5 g
M183825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18385G Supplier TCI America Supplier No. M18385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 63012-97-5
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.189
MDL Number MFCD03701635
InChI Key OQVAOEIMSKZGAL-UHFFFAOYSA-N
PubChem CID 526618
IUPAC Name 2-methyl-3-methylsulfanylfuran
SMILES CC1=C(C=CO1)SC

Specifications

Color Yellow
Boiling Point 132°C
Quantity 5 g
UN Number 1993
Formula Weight 128.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-(methylthio)furan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.