Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitrobenzoic acid, 99%, Thermo Scientific™

Catalog No. AAA1469430
Change view
Click to view available options
Quantity:
100 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1469430 250 g
AAA1469422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1469430 Supplier Thermo Scientific Chemicals Supplier No. A1469430

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1975-50-4
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007160
InChI Key YPQAFWHSMWWPLX-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzoic acid, 3-nitro-o-toluic acid, benzoic acid, 2-methyl-3-nitro, 2-methyl-3-nitrobenzoicacid, unii-0si17d349b, 3-nitro-2-methyl benzoic acid, 3-nitro-2-methylbenzoic acid, 2-methyl-3-nitro-benzoic acid, benzoic acid,2-methyl-3-nitro, pubchem4522
PubChem CID 16096
SMILES CC1=C(C=CC=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point 182°C to 186°C
Quantity 250 g
Beilstein 2050096
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Methyl-3-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-826-4
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.