Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M089225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M089225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M089225G Supplier TCI America Supplier No. M089225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1975-50-4
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007160
InChI Key YPQAFWHSMWWPLX-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzoic acid, 3-nitro-o-toluic acid, benzoic acid, 2-methyl-3-nitro, 2-methyl-3-nitrobenzoicacid, unii-0si17d349b, 3-nitro-2-methyl benzoic acid, 3-nitro-2-methylbenzoic acid, 2-methyl-3-nitro-benzoic acid, benzoic acid,2-methyl-3-nitro, pubchem4522
PubChem CID 16096
SMILES CC1=C(C=CC=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point 184°C
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitrobenzoic Acid
EINECSNumber (3)-4284
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.