Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-nitropyridine, 97%, Thermo Scientific™

Catalog No. AC453690010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453690010 1 g
AC453690050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453690010 Supplier Thermo Scientific Chemicals Supplier No. 453690010
Add to Cart

Chemical Identifiers

CAS 18699-87-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD04114137
InChI Key CCFGTKQIRWHYTB-UHFFFAOYSA-N
Synonym 2-methyl-3-nitro-pyridine, 3-nitro-2-picoline, 3-nitropicoline, 3-nitro-2-methylpyridine, pyridine, 2-methylnitro, pyridine, 2-methyl-3-nitro-9ci, nitropicoline, zlchem 137, acmc-209ep0, pyridine, 2-methyl-3-nitro
PubChem CID 329360
IUPAC Name 2-methyl-3-nitropyridine
SMILES CC1=NC=CC=C1[N+]([O-])=O

Specifications

Boiling Point 104.0°C (15.0 mmHg)
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 138.13
Percent Purity 97%
Chemical Name or Material 2-Methyl-3-nitropyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.