Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-octanol 98.0+%, TCI America™
SDP

Catalog No. M053510ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M053510ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M053510ML Supplier TCI America Supplier No. M053510ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26533-34-6
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00021946
InChI Key DIVBBSLQUDHECU-UHFFFAOYNA-N
PubChem CID 98464
IUPAC Name 2-methyloctan-3-ol
SMILES CCCCCC(O)C(C)C

Specifications

Color Colorless
Boiling Point 189°C
Quantity 10 mL
Formula Weight 144.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-octanol
EINECSNumber (2)-0217
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.