Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-pentanone 98.0+%, TCI America™
SDP

Catalog No. M124925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M124925ML 25 mL
M12495ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M124925ML Supplier TCI America Supplier No. M124925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 565-69-5
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009314
InChI Key HYTRYEXINDDXJK-UHFFFAOYSA-N
Synonym 2-methyl-3-pentanone, ethyl isopropyl ketone, 3-pentanone, 2-methyl, 4-methyl-3-pentanone, isopropyl ethyl ketone, 2-methyl-3-pentanal, iso-c3h7coc2h5, acmc-209lte, ksc272m0l
PubChem CID 11265
IUPAC Name 2-methylpentan-3-one
SMILES CCC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
UN Number 1224
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-pentanone
EINECSNumber (2)-2475
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.