Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-phenyl-1-propene, MP Biomedicals

Catalog No. ICN225257 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225257 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225257 Supplier MP Biomedicals, Inc Supplier No. 0522525705

Chemical Identifiers

CAS 3290-53-7
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
InChI Key MXTNFIYGTWARIN-UHFFFAOYSA-N
Synonym 2-methyl-3-phenyl-1-propene, methallylbenzene, benzene, 2-methyl-2-propenyl, benzene, 2-methylallyl, 2-methylprop-2-en-1-yl benzene, 2-methyl-2-propenyl benzene, methallyl benzene, 2-methylallylbenzene, 2-benzyl-1-propene, 2-methylallyl benzene
PubChem CID 18687
IUPAC Name 2-methylprop-2-enylbenzene
SMILES CC(=C)CC1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 132.21
Chemical Name or Material 2-Methyl-3-Phenyl-1-Propene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.