Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-3-propylpyrazine 95.0+%, TCI America™
SDP

Catalog No. M084125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M084125ML 25 mL
M08415ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M084125ML Supplier TCI America Supplier No. M084125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15986-80-8
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00010096
InChI Key XAWKNALRUSOTOY-UHFFFAOYSA-N
Synonym 2-methyl-3-n-propylpyrazine, pyrazine, 2-methyl-3-propyl, 2-propyl-3-methylpyrazine, 3-propyl-2-methylpyrazine, 2-methyl-3-propyl-pyrazine, 3-propyl-2-methyl pyrazine, 2-methyl-3-propylpyrazin, methylpropylpyrazine, acmc-209dk8, chembl95176
PubChem CID 85224
IUPAC Name 2-methyl-3-propylpyrazine
SMILES CCCC1=NC=CN=C1C

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 mL
Formula Weight 136.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-propylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.