Learn More
Medchemexpress LLC 2-methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b]benzodiazepine | 132539-06-1 | MFCD00866702 | 100.0% | 312.43 | C17H20N4S | 5 MG
SDP
Catalog No. 5000221618
Encompass_Preferred
Analytical reference standard of olanzapine for research and analytical applications. Olanzapine is an atypical antipsychotic that acts as a monoaminergic antagonist with high affinity for multiple serotonin and dopamine receptors. This standard is intended for qualitative, quantitative, and methodological workflows including HPLC, GC, and MS and is supplied with a certificate of analysis.
  • High purity suitable for analytical use.
  • Solid, light yellow to yellow appearance.
  • Intended for HPLC, GC, and MS applications.
  • Supplied in multiple small pack sizes for reference work.
  • Certificate of analysis included for storage and handling instructions.
Catalog No. 50-002-21618 Supplier Medchemexpress LLC Supplier No. HY14541R5MG
Analytical reference standard of olanzapine for research and analytical applications. Olanzapine is an atypical antipsychotic that acts as a monoaminergic antagonist with high affinity for multiple serotonin and dopamine receptors. This standard is intended for qualitative, quantitative, and methodological workflows including HPLC, GC, and MS and is supplied with a certificate of analysis.
  • High purity suitable for analytical use.
  • Solid, light yellow to yellow appearance.
  • Intended for HPLC, GC, and MS applications.
  • Supplied in multiple small pack sizes for reference work.
  • Certificate of analysis included for storage and handling instructions.