Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4(5)-nitroimidazole, TCI America™
SDP

Catalog No. M0922500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0922500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M0922500G Supplier TCI America Supplier No. M0922500G

missing translation for 'validMainframeMsg'

Specifications

CAS 696-23-1
Melting Point 253°C
Color Yellow
Molecular Formula C4H5N3O2
Quantity 500 g
MDL Number MFCD00005191
Synonym 2-methyl-5-nitroimidazole, 2-methyl-4-nitroimidazole, 2-methyl-4-nitro-1h-imidazole, menidazole, 1h-imidazole, 2-methyl-4-nitro, 1h-imidazole, 2-methyl-5-nitro, 2-methyl-4 5-nitroimidazole, imidazole, 2-methyl-4-nitro, unii-24ag2ww15w, imidazole, 2-methyl-5-nitro
InChI Key FFYTTYVSDVWNMY-UHFFFAOYSA-N
SMILES CC1=NC=C(N1)[N+]([O-])=O
IUPAC Name 2-methyl-5-nitro-1H-imidazole
Molecular Weight (g/mol) 127.10
PubChem CID 12760
Formula Weight 127.10
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4(5)-nitroimidazole
Show More Show Less
RTECSNumber NI7550000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.