Learn More
Medchemexpress LLC 2-Methyl-4-pentenoic Acid | 1575-74-2 | 99.8% | 114.14 | 1 ML
SDP
Catalog No. 5000451958
Encompass_Preferred
2-Methyl-4-pentenoic Acid is an organic acid that can be used as a precursor for synthesizing other compounds. Additionally, changes in the level of 2-Methyl-4-pentenoic Acid may be associated with the susceptibility to schizophrenia and type 2 diabetes. This product is for research use only.
  • Liquid appearance (density: 0.956 g/cm3)
  • Colorless to light yellow
  • SMILES: C=CCC(C)C(O)=O
  • Classified as ketones, aldehydes, and acids
  • Initial source: Plants other families
Catalog No. 50-004-51958 Supplier Medchemexpress LLC Supplier No. HYW01292210MM/1ML
2-Methyl-4-pentenoic Acid is an organic acid that can be used as a precursor for synthesizing other compounds. Additionally, changes in the level of 2-Methyl-4-pentenoic Acid may be associated with the susceptibility to schizophrenia and type 2 diabetes. This product is for research use only.
  • Liquid appearance (density: 0.956 g/cm3)
  • Colorless to light yellow
  • SMILES: C=CCC(C)C(O)=O
  • Classified as ketones, aldehydes, and acids
  • Initial source: Plants other families