Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-pentenoic Acid 98.0+%, TCI America™
SDP

Catalog No. M114310ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M114310ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M114310ML Supplier TCI America Supplier No. M114310ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1575-74-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00015570
InChI Key HVRZYSHVZOELOH-UHFFFAOYSA-N
Synonym 2-methyl-4-pentenoic acid, 4-pentenoic acid, 2-methyl, 2-methylpent-4-en-1-oic acid, fema no. 3511, 2-methyl-4-pentenoicacid, methylallylacetic acid, acmc-209dg9, 2-methyl-pent-4-enoic acid, hvrzyshvzoeloh-uhfffaoysa, 2-methyl-4-pentenoic acid, fg
PubChem CID 549519
IUPAC Name 2-methylpent-4-enoic acid
SMILES CC(CC=C)C(=O)O

Specifications

Boiling Point 104°C
Quantity 10 mL
UN Number 3265
Formula Weight 114.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methyl-4-pentenoic Acid
EINECSNumber (2)-0609
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.