Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-phenylthiazole 98.0+%, TCI America™
SDP

Catalog No. M28925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28925G 5 g
M28921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28925G Supplier TCI America Supplier No. M28925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1826-16-0
Molecular Formula C10H9NS
Molecular Weight (g/mol) 175.249
MDL Number MFCD00489998
InChI Key XROORURTAQOYLW-UHFFFAOYSA-N
Synonym 2-methyl-4-phenylthiazole, 2-methyl-4-phenyl-thiazole, thiazole, 2-methyl-4-phenyl, thiazole,2-methyl-4-phenyl, #
PubChem CID 584377
IUPAC Name 2-methyl-4-phenyl-1,3-thiazole
SMILES CC1=NC(=CS1)C2=CC=CC=C2

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 284°C
Quantity 5 g
Formula Weight 175.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-4-phenylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.