Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Catalog No. AAH6177903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6177903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6177903 Supplier Thermo Scientific Chemicals Supplier No. H6177903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67169-22-6
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.154
MDL Number MFCD03701068
InChI Key PAXQXJDYVORMOO-UHFFFAOYSA-N
Synonym 2-methyl-4-trifluoromethyl aniline, 4-amino-3-methylbenzotrifluoride, benzenamine, 2-methyl-4-trifluoromethyl, 2-amino-5-trifluoromethyl toluene, pubchem4492, 2-amino-5-trifluoromethyltoluene, 2-methyl-4 trifluoromethyl benzenamine, 2-methyl4-trifluoromethyl benzenamine, 2-methyl-4-trifluoromethyl benzenamine
PubChem CID 13750437
IUPAC Name 2-methyl-4-(trifluoromethyl)aniline
SMILES CC1=C(C=CC(=C1)C(F)(F)F)N

Specifications

Color White
Quantity 1 g
UN Number UN2811
Formula Weight 175.15
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Methyl-4-(trifluoromethyl)aniline
Hazard Category Danger
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.