Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-4-undecanone 97.0+%, TCI America™
SDP

Catalog No. I025025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I025025G 25 g
I02501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I025025G Supplier TCI America Supplier No. I025025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19594-40-2
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.323
MDL Number MFCD00027319
InChI Key IEBAJFDSHJYDCK-UHFFFAOYSA-N
Synonym Heptyl Isobutyl Ketone
PubChem CID 246924
IUPAC Name 2-methylundecan-4-one
SMILES CCCCCCCC(=O)CC(C)C

Specifications

Color Yellow
Boiling Point 124°C
Quantity 25 g
Formula Weight 184.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-4-undecanone
EINECSNumber (2)-2475
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.