Learn More
TARGETMOL CHEMICALS INC 2-Methyl-5-HT 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1007525MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 2-Methyl-5-HT (2-Methyl-5-hydroxytryptamine) is a potent, selective 5-HT3 receptor agonist exhibiting anti-depressive-like effects. Purity 95.64%
Catalog No. 50-288-0351