Learn More
Medchemexpress LLC HY-19358 5mg Medchemexpress, 2-Methyl-5-HT CAS:78263-90-8 Purity:>98%
SDP
Catalog No. 501970024
Encompass_Preferred
Medchemexpress, HY-19358 5mg 2-Methyl-5-HT CAS:78263-90-8 2-Methyl-5-HT (2-Methyl-5-hydroxytryptamine) is a potent and selective 5-HT3 receptor agonist. 2-Methyl-5-HT is shown to display anti-depressive-like effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0024 Supplier Medchemexpress LLC Supplier No. HY193585MG
Medchemexpress, HY-19358 5mg 2-Methyl-5-HT CAS:78263-90-8 2-Methyl-5-HT (2-Methyl-5-hydroxytryptamine) is a potent and selective 5-HT3 receptor agonist. 2-Methyl-5-HT is shown to display anti-depressive-like effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.