Learn More
Medchemexpress LLC 2-Methyl-5-HT | 78263-90-8 | 98.9% | 190.24 | 25 MG
SDP
Catalog No. 5000384411
Encompass_Preferred
2-Methyl-5-HT (2-Methyl-5-hydroxytryptamine) is a potent and selective 5-HT3 receptor agonist. It has been shown to display anti-depressive-like effects.
  • Potent and selective 5-HT3 receptor agonist.
  • Shown to display anti-depressive-like effects.
Catalog No. 50-003-84411 Supplier Medchemexpress LLC Supplier No. 25MGHY19358
2-Methyl-5-HT (2-Methyl-5-hydroxytryptamine) is a potent and selective 5-HT3 receptor agonist. It has been shown to display anti-depressive-like effects.
  • Potent and selective 5-HT3 receptor agonist.
  • Shown to display anti-depressive-like effects.