Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals

Catalog No. AC248790250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC248790250 25 g
AC248790010 1 g
AC248790050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC248790250 Supplier Thermo Scientific Chemicals Supplier No. 248790250
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2051-53-8 | C10H15N | 149.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2051-53-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00044503
InChI Key YKWALWNGEXPARQ-UHFFFAOYSA-N
IUPAC Name 2-methyl-5-(propan-2-yl)aniline
SMILES CC(C)C1=CC=C(C)C(N)=C1

Specifications

Alpha Vector METHYLISOPROPYLANILINE
Color Brown to Colorless
Density 0.95
Infrared Spectrum Conforms
Packaging glass bottle
Refractive Index 1.5350 to 1.5450 (20°C, 589 nm)
Quantity 25 g
Sensitivity Air sensitive
Formula Weight 149.23
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-isopropylaniline
Show More Show Less
Hazard Category GHS07: Harmful
ShelfLife 3 years
storageNote1 normal conditions

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.