Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-nitrophenol, 97%, Thermo Scientific™

Catalog No. AAB2047206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2047206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2047206 Supplier Thermo Scientific Chemicals Supplier No. B2047206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5428-54-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00043909
InChI Key UMFDLIXUUJMPSI-UHFFFAOYSA-N
Synonym 5-nitro-o-cresol, 5-nitro-2-cresol, phenol, 2-methyl-5-nitro, 2-hydroxy-4-nitrotoluene, o-cresol, 5-nitro, 5-nitro-2-methylphenol, 2-methy-5-nitrophenol, 2-methyl-5-nitro phenol, 2-methyl-5-nitro-phenol, 4-nitro-2-hydroxytoluene
PubChem CID 93576
IUPAC Name 2-methyl-5-nitrophenol
SMILES CC1=CC=C(C=C1O)[N+]([O-])=O

Specifications

Melting Point 116°C to 120°C
Boiling Point 180°C (15 mmHg)
Odor Phenol-like
Quantity 5 g
UN Number UN2446
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-nitrophenol
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID
EINECSNumber 226-580-7
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.