Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-5-nitropyridine, 95%

Catalog No. AAH6175006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6175006 5 g
AAH6175014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6175006 Supplier Thermo Scientific Chemicals Supplier No. H6175006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 21203-68-9 | C6H6N2O2 | 138.126 g/mol

2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21203-68-9
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD04114179
InChI Key USZINSZJSVMICC-UHFFFAOYSA-N
Synonym 5-nitro-2-picoline, pyridine, 2-methyl-5-nitro, 5-nitro-2-methylpyridine, 2-methyl-5-nitro-pyridine, pubchem6699, acmc-1cewl, 2-methyl-5-nitro pyridine, ksc494i9p, abbypharma ap-15-5036
PubChem CID 2794552
IUPAC Name 2-methyl-5-nitropyridine
SMILES CC1=NC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 138.12
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Methyl-5-nitropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.