Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-6-nitrobenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. M14381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14381G 1 g
M14385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14381G Supplier TCI America Supplier No. M14381G

Chemical Identifiers

CAS 66232-57-3
Molecular Formula C8H6ClNO3
Molecular Weight (g/mol) 199.59
MDL Number MFCD06797116
InChI Key ZMAJSTDJAIVHGZ-UHFFFAOYSA-N
Synonym 6-Nitro-o-toluoyl Chloride
PubChem CID 12819896
IUPAC Name 2-methyl-6-nitrobenzoyl chloride
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])C(=O)Cl

Specifications

Melting Point 42°C
Quantity 1 g
UN Number 3261
Formula Weight 199.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-6-nitrobenzoyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.