Learn More
Electron Microscopy Sciences 2-Methyl Butane, Reagent 450 ML

Supplier: Electron Microscopy Sciences 18550
(Isopropentane)
CAS #78-78-4 CH₂H₅CH(CH₃)₂ F.W. 72.15 g/mol
Melting point: –158.5 °C (–253 °F)
Boiling point: 27-31°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.