Learn More
Medchemexpress LLC 2-methyl-N-([1-(4-phenylpiperazin-1-yl)cyclohexyl]methyl)-1,2,3,4-tetrahydroisoquinoline-5-carboxami | 1428327-35-8 | MFCD34676648 | 98.2% | 446.63 | C28H38N4O | 5MG
SDP
Catalog No. 5000237944
Encompass_Preferred
JNJ-42253432 is a centrally penetrant, high-affinity, orally active P2X7 receptor antagonist used as a research tool to study P2X7-mediated neuroinflammation and brain receptor occupancy. It exhibits high potency in preclinical assays and has documented in vivo brain occupancy at low systemic doses.
Catalog No. 50-002-37944 Supplier Medchemexpress LLC Supplier No. HY1234815MG
JNJ-42253432 is a centrally penetrant, high-affinity, orally active P2X7 receptor antagonist used as a research tool to study P2X7-mediated neuroinflammation and brain receptor occupancy. It exhibits high potency in preclinical assays and has documented in vivo brain occupancy at low systemic doses.