Learn More
Medchemexpress LLC 2-Methyl-N,N-bis(methyl-d3)-3-(10H-phenothiazin-10-yl)propan-1-amine | 1346603-88-0 | 99.6% | 1 MG
SDP
Catalog No. 5000342486
Encompass_Preferred
Alimemazine-d6 is a deuterium-labeled form of Alimemazine, an antihistamine. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Research use only compound
  • Functions as a tracer in various studies
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeled compound
  • Related to isotope-labeled compounds and internal standards for clinical mass spectrometry
  • Relevant for neurological, eye or ear disease, and inflammation or immune system disease research
Catalog No. 50-003-42486 Supplier Medchemexpress LLC Supplier No. HY12752S1MG
Alimemazine-d6 is a deuterium-labeled form of Alimemazine, an antihistamine. It is intended for research use only and can serve as a tracer or an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Research use only compound
  • Functions as a tracer in various studies
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium labeled compound
  • Related to isotope-labeled compounds and internal standards for clinical mass spectrometry
  • Relevant for neurological, eye or ear disease, and inflammation or immune system disease research