Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methyl-N,N-bis(methyl-d3)-3-(10H-phenothiazin-10-yl)propan-1-amine | 1346603-88-0 | MFCD28900690 | 99.6% | 304.48 g/mol | C18H16D6N2S | 5 MG
SDP

Catalog No. 5000264610
Encompass_Preferred

Alimemazine-d6 is a deuterium-labeled analogue of alimemazine supplied as a research-grade analytical standard. It is intended for laboratory use as an internal standard or tracer in quantitative analyses and supports accurate measurement in spectrometric and chromatographic workflows.

  • High isotopic labeling for improved quantitation in MS and NMR.
  • Chemical purity approximately 99.6%.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • Solid form, white to off-white appearance.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Molecular weight 304.48 g/mol; formula C18H16D6N2S.

Catalog No. 50-002-64610 Supplier Medchemexpress LLC Supplier No. HY12752S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Alimemazine-d6 is a deuterium-labeled analogue of alimemazine supplied as a research-grade analytical standard. It is intended for laboratory use as an internal standard or tracer in quantitative analyses and supports accurate measurement in spectrometric and chromatographic workflows.

  • High isotopic labeling for improved quantitation in MS and NMR.
  • Chemical purity approximately 99.6%.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS.
  • Solid form, white to off-white appearance.
  • Recommended storage: 4°C, protect from light; in solvent: -80°C for 6 months, -20°C for 1 month.
  • Molecular weight 304.48 g/mol; formula C18H16D6N2S.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.