Learn More
Medchemexpress LLC 2-Methylacetophenone | 577-16-2 | 99.4% | 134.18 | 1 ML
SDP
Catalog No. 5000454834
Encompass_Preferred
2-Methylacetophenone (O-Methylacetophenone) is a xanthine dehydrogenase (XDH) inhibitor that competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. This product is suitable for the study of hyperuricemia.
  • Inhibits xanthine dehydrogenase (XDH)
  • Blocks conversion of xanthine to uric acid
  • Can be used for hyperuricemia studies
  • Appears as a colorless to light yellow liquid
  • Intended for research and analytical applications
Catalog No. 50-004-54834 Supplier Medchemexpress LLC Supplier No. HYW01265810MM/1ML
2-Methylacetophenone (O-Methylacetophenone) is a xanthine dehydrogenase (XDH) inhibitor that competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. This product is suitable for the study of hyperuricemia.
  • Inhibits xanthine dehydrogenase (XDH)
  • Blocks conversion of xanthine to uric acid
  • Can be used for hyperuricemia studies
  • Appears as a colorless to light yellow liquid
  • Intended for research and analytical applications