Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2'-Methylacetophenone

Catalog No. M2659325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2659325G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2659325G Supplier Sigma Aldrich Supplier No. M2659325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2'-Methylacetophenone

Specifications

CAS 577-16-2
Density 1.026 g/mL (at 25°C (literature))
Boiling Point 214°C (lit.)
Molecular Formula C9H10O
Refractive Index n20/D 1.5318 (literature)
Linear Formula CH3C6H4COCH3
MDL Number MFCD00008734
Quantity 1 g
Synonym 1-(2-Methylphenyl)ethanone; 1-(2-Tolyl)ethanone; 2-Acetyltoluene; o-Methylacetophenone; Methyl o-tolyl ketone
Molecular Weight (g/mol) 134.18
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.