Learn More
Medchemexpress LLC 2-Methylacetophenone (O-Methylacetophenone) | 577-16-2 | 99.4% | 134.18 | 10 G
SDP
Catalog No. 5000454865
Encompass_Preferred
2-Methylacetophenone (O-Methylacetophenone) is an analytical standard and a xanthine dehydrogenase (XDH) inhibitor. It competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. This compound is intended for research and analytical applications, and can be used for the study of hyperuricemia.
  • Xanthine dehydrogenase (XDH) inhibitor.
  • Competitively binds to the XDH active site.
  • Blocks the pathway for xanthine to be converted to uric acid.
  • Can be used for the study of hyperuricemia.
  • Intended for research and analytical applications.
Catalog No. 50-004-54865 Supplier Medchemexpress LLC Supplier No. HYW01265810G
2-Methylacetophenone (O-Methylacetophenone) is an analytical standard and a xanthine dehydrogenase (XDH) inhibitor. It competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. This compound is intended for research and analytical applications, and can be used for the study of hyperuricemia.
  • Xanthine dehydrogenase (XDH) inhibitor.
  • Competitively binds to the XDH active site.
  • Blocks the pathway for xanthine to be converted to uric acid.
  • Can be used for the study of hyperuricemia.
  • Intended for research and analytical applications.