Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Methylacetophenone (O-Methylacetophenone) | 577-16-2 | 99.6% | 134.18 | 5 G
SDP

Catalog No. 5000455117
Encompass_Preferred

2-Methylacetophenone (O-Methylacetophenone) is a xanthine dehydrogenase (XDH) inhibitor. It competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. It can be used for the study of hyperuricemia.

  • Molecular weight: 134.18
  • Formula: C9H10O
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage: -20°C for 3 years in pure form.
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-55117 Supplier Medchemexpress LLC Supplier No. HYW0126585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Methylacetophenone (O-Methylacetophenone) is a xanthine dehydrogenase (XDH) inhibitor. It competitively binds to the XDH active site, blocking the pathway for xanthine to be converted to uric acid. It can be used for the study of hyperuricemia.

  • Molecular weight: 134.18
  • Formula: C9H10O
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage: -20°C for 3 years in pure form.
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.