Learn More
Strem, An Ascensus Company CAS# 1581285-85-9. 2g. (2'-Methylamino-1,1'-biphenyl-2-yl)methanesulfonatopalladium(II) dimer, min. 98%.
SDP
Supplier:  Strem, An Ascensus Company 4615532G
Encompass
CAS# 1581285-85-9. 2g. (2'-Methylamino-1,1'-biphenyl-2-yl)methanesulfonatopalladium(II) dimer, min. 98%. . Molecular Weight: 767.52. Molecular Formula: C28H30N2O6Pd2S2. Color/form: pale yellow to light gray pwdr. Strem# 46-1553. http://www.strem.com/catalog/v/46-1553/
Catalog No. NC0896812